Learn More
Abnova™ MIR19q13.42/CEN19p FISH Probe
Catalog No. 89337309 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
FISH-Ce
Catalog No. For Use With (Application)
89-337-309 FISH-Ce
1 options
Catalog No. 89-337-309 Supplier Abnova™ Supplier No. FG0250
Add to Cart
Add to Cart
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Specifications
For Use With (Application) FISH-Ce