Learn More
Medchemexpress LLC MIRA-1 | 72835-26-8 | 99.8% | 183.16 | 50 MG
SDP
Catalog No. 5000344293
Encompass_Preferred
MIRA-1 is a maleimide analogue that can induce apoptosis in mutant p53 cells via restoration of p53-dependent transcriptional transactivation. It exhibits anticancer activity.
  • Induces apoptosis in mutant p53 cells.
  • Restores p53-dependent transcriptional transactivation.
  • Possesses anticancer activity.
  • Suppresses the growth of Saos-2-His273 cells expressing mutant p53.
  • Inhibits cell growth in a mutant p53-dependent manner.
Catalog No. 50-003-44293 Supplier Medchemexpress LLC Supplier No. HY10863910MG
MIRA-1 is a maleimide analogue that can induce apoptosis in mutant p53 cells via restoration of p53-dependent transcriptional transactivation. It exhibits anticancer activity.
  • Induces apoptosis in mutant p53 cells.
  • Restores p53-dependent transcriptional transactivation.
  • Possesses anticancer activity.
  • Suppresses the growth of Saos-2-His273 cells expressing mutant p53.
  • Inhibits cell growth in a mutant p53-dependent manner.