Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mira-1 | 72835-26-8 | 99.8% | 183.16 g/mol | C8H9NO4 | 5 MG
SDP

Catalog No. 5000264267
Encompass_Preferred

MIRA-1 (NSC 19630) is a maleimide-derived small-molecule research compound that restores wild-type conformation and transcriptional activity to mutant p53, triggering p53-dependent transcriptional transactivation and apoptosis in mutant p53 cancer cells. It has reported anticancer activity and an IC50 of approximately 10 μM against Saos-2 His273 cells. Provided as a high-purity reagent for in vitro and in vivo studies.

  • Molecular formula: C8H9NO4.
  • Molecular weight: 183.16 g/mol.
  • Purity: 99.76%.
  • Solubility: DMSO 100 mg/mL; in vivo vehicles soluble ≥2.5 mg/mL.
  • Suggested storage: powder -20°C for long term; in solvent -80°C recommended.
  • Reported activity: induces p53-dependent transcriptional transactivation and apoptosis.

Catalog No. 50-002-64267 Supplier Medchemexpress LLC Supplier No. HY1086395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MIRA-1 (NSC 19630) is a maleimide-derived small-molecule research compound that restores wild-type conformation and transcriptional activity to mutant p53, triggering p53-dependent transcriptional transactivation and apoptosis in mutant p53 cancer cells. It has reported anticancer activity and an IC50 of approximately 10 μM against Saos-2 His273 cells. Provided as a high-purity reagent for in vitro and in vivo studies.

  • Molecular formula: C8H9NO4.
  • Molecular weight: 183.16 g/mol.
  • Purity: 99.76%.
  • Solubility: DMSO 100 mg/mL; in vivo vehicles soluble ≥2.5 mg/mL.
  • Suggested storage: powder -20°C for long term; in solvent -80°C recommended.
  • Reported activity: induces p53-dependent transcriptional transactivation and apoptosis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.