Learn More
Thermo Scientific MIRA MIST NEBULIZER
Supplier:  Thermo Scientific 842312051471
Encompass
Mira Mist Nebuliser; HF resistant, parallel path nebulizer intended for the analysis of; samples containing HF acid and/or samples containing high levels of; dissolved solids.; The unique parallel path design provides enhanced resistance to; blockages when running samples containing; fine particulates and dissolved solids.; The nebulizer design enables liquid aspiration over a variable flow; rate of 0.2-2.5 ml/min and is compatible with all iCAP 6000 Series; Spectrometers
Catalog No. NC9532943