Learn More
Cayman Chemical Mirabegron Impurity 6, Purity: ≥95%, CAS: 391901-45-4, 500 mg
SDP
Catalog No. 502912385
Encompass_Preferred
A metabolite of mirabegron; formed via hydrolysis of mirabegron by BChE; an intermediate in the synthesis of mirabegron
Catalog No. 50-291-2385 Supplier Cayman Chemical Supplier No. 36315500 MG
A metabolite of mirabegron; formed via hydrolysis of mirabegron by BChE; an intermediate in the synthesis of mirabegron