Learn More
Perkin Elmer US LLC Miracle ATR Top Plate - Diamond/ZnSe
Catalog No. 502112730
Encompass_Preferred
The Miracle ATR Sampling Accessory is used to analyze of solids, liquids, pastes, gels, and intractable materials. In its most popular configuration, it acts as a single reflection ATR accessory with high IR throughput, which makes it ideal for sample identification and QA/QC applications. The easily changeable crystal plate design provides an analysis of a broad spectrum of sample types, while ensuring a constant sampling pathlength.
Catalog No. 50-211-2730 Supplier Perkin Elmer US LLC Supplier No. L1272313
Add to Cart
Add to Cart
Request a Quote
The Miracle ATR Sampling Accessory is used to analyze of solids, liquids, pastes, gels, and intractable materials. In its most popular configuration, it acts as a single reflection ATR accessory with high IR throughput, which makes it ideal for sample identification and QA/QC applications. The easily changeable crystal plate design provides an analysis of a broad spectrum of sample types, while ensuring a constant sampling pathlength.