Learn More
Perkin Elmer US LLC Miracle ATR/10 Reflection HATR USB Temperature Controller (220V, UK)
Catalog No. 502112725
Encompass_Preferred
The Horizontal Attenuated Total Reflectance (HATR) features a constant and reproducible effective path length and is well suited for both qualitative and quantitative applications. The HATR may be configured with flat or trough plates and with this controller, has temperature guidance. This accessory is compatible with the Spectrum Two and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-2725 Supplier Perkin Elmer US LLC Supplier No. L1272317
The Horizontal Attenuated Total Reflectance (HATR) features a constant and reproducible effective path length and is well suited for both qualitative and quantitative applications. The HATR may be configured with flat or trough plates and with this controller, has temperature guidance. This accessory is compatible with the Spectrum Two and Spectrum 3/Frontier spectrometers.