Learn More
Perkin Elmer US LLC MIRacle Heated Crystal Plate - ZnSe, 130°C
Catalog No. 502112726
Encompass_Preferred
The MIRacle ATR accessory analyzes solids, liquids, pastes, gels, and intractable materials. In its most popular configuration, it acts as a single reflection ATR accessory with high IR throughput, which makes it ideal for sample identification and QA/QC applications. The easily changeable crystal plate design provides an analysis of a broad spectrum of sample types, while ensuring a constant sampling path length.
Catalog No. 50-211-2726 Supplier Perkin Elmer US LLC Supplier No. L1272315
The MIRacle ATR accessory analyzes solids, liquids, pastes, gels, and intractable materials. In its most popular configuration, it acts as a single reflection ATR accessory with high IR throughput, which makes it ideal for sample identification and QA/QC applications. The easily changeable crystal plate design provides an analysis of a broad spectrum of sample types, while ensuring a constant sampling path length.