Learn More
Medchemexpress LLC HY-19719 5mg Medchemexpress, Miransertib CAS:1313881-70-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY197195MG
Encompass_Preferred
Medchemexpress, HY-19719 5mg Miransertib CAS:1313881-70-7 Miransertib (ARQ-092) is a potent, orally active, selective and allosteric Akt inhibitor with IC50s of 2.7 nM, 14 nM and 8.1 nM for Akt1, Akt2, Akt3, respectively. Miransertib is also a potent the AKT1-E17K mutant protein inhibitor and has the potential for PI3K/AKT-driven tumors and Proteus syndrome research. Miransertib is effective against Leishmania[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9289
 Restricted Item