Learn More
Medchemexpress LLC Miransertib hydrochloride (ARQ-092 hydrochloride) | 1313883-00-9 | 99.7% | 468.98 | 100 MG
SDP
Catalog No. 5000401341
Encompass_Preferred
Miransertib hydrochloride (ARQ-092 hydrochloride) is a potent, orally active, selective, and allosteric Akt inhibitor. It exhibits specific IC50s for Akt1, Akt2, and Akt3, and is a potent inhibitor of the AKT1-E17K mutant protein. This compound is considered for research into PI3K/AKT-driven tumors, Proteus syndrome, and is effective against Leishmania.
  • Potent, orally active, selective, and allosteric Akt inhibitor.
  • Exhibits specific IC50s for Akt1, Akt2, and Akt3.
  • Inhibits the AKT1-E17K mutant protein.
  • Suitable for research in PI3K/AKT-driven tumors and Proteus syndrome.
  • Effective against Leishmania.
Catalog No. 50-004-01341 Supplier Medchemexpress LLC Supplier No. HY19719A100MG
Miransertib hydrochloride (ARQ-092 hydrochloride) is a potent, orally active, selective, and allosteric Akt inhibitor. It exhibits specific IC50s for Akt1, Akt2, and Akt3, and is a potent inhibitor of the AKT1-E17K mutant protein. This compound is considered for research into PI3K/AKT-driven tumors, Proteus syndrome, and is effective against Leishmania.
  • Potent, orally active, selective, and allosteric Akt inhibitor.
  • Exhibits specific IC50s for Akt1, Akt2, and Akt3.
  • Inhibits the AKT1-E17K mutant protein.
  • Suitable for research in PI3K/AKT-driven tumors and Proteus syndrome.
  • Effective against Leishmania.