Learn More
Medchemexpress LLC HY-10254 25mg , Mirdametinib CAS:391210-10-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1025425MG
Encompass_Preferred
HY-10254 25mg Mirdametinib CAS: 391210-10-9 Mirdametinib (PD0325901) is an orally active, selective and non-ATP-competitive MEK inhibitor with an IC50 of 0.33 nM. Mirdametinib exhibits a Kiapp of 1 nM against activated MEK1 and MEK2. Mirdametinib suppresses the expression of p-ERK1/2 and induces apoptosis. Mirdametinib has anti-cancer activity for a broad spectrum of human tumor xenografts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9299