Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc Mirex 100 µg/mL in MeOH 1ml
SDP

Catalog No. NC9015645
Encompass

Mirex . 1 mL. 100 g/mL in MeOH. 2385-85-5

Catalog No. NC9015645 Supplier AccuStandard, Inc Supplier No. P066S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mirex . 1 mL. 100 g/mL in MeOH. 2385-85-5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.