Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mirificin | 103654-50-8 | 5 MG
SDP

Catalog No. 5000421878
Encompass_Preferred

Mirificin is an isoflavone found in Puerariae Lobatae Radix. It acts as a tyrosinase (TYR) inhibitor with an IC50 of 12.66 μM.

  • Purity of 99.66%
  • Also known as Puerarin apioside
  • Inhibits tyrosinase with an IC50 of 12.66 μM
  • Molecular weight of 548.49
  • Chemical formula C26H28O13
  • Solid appearance
  • Color ranges from white to light yellow
  • Classified as flavonoid, isoflavone, phenol, and polyphenol
  • Initially sourced from plants

Catalog No. 50-004-21878 Supplier Medchemexpress LLC Supplier No. HYN21345MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mirificin is an isoflavone found in Puerariae Lobatae Radix. It acts as a tyrosinase (TYR) inhibitor with an IC50 of 12.66 μM.

  • Purity of 99.66%
  • Also known as Puerarin apioside
  • Inhibits tyrosinase with an IC50 of 12.66 μM
  • Molecular weight of 548.49
  • Chemical formula C26H28O13
  • Solid appearance
  • Color ranges from white to light yellow
  • Classified as flavonoid, isoflavone, phenol, and polyphenol
  • Initially sourced from plants

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.