Learn More
Selleck Chemical LLC Mirin 10mg 1198097-97-0
SDP
Supplier:  Selleck Chemical LLC S809610MG
Encompass_Preferred
Mirin is a potent Mre11¿Rad50¿Nbs1 (MRN) complex inhibitor, and inhibits Mre11-associated exonuclease activity. Mirin inhibits MRN-dependent activation of ATM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5645