Learn More
Medchemexpress LLC Mirin | 299953-00-7 | MFCD05885480 | 99.2% | 220.25 | C10H8N2O2S | 50 MG
SDP
Catalog No. 5000226682
Encompass_Preferred
(E/Z)-Mirin is a research-grade small-molecule inhibitor of the Mre11-Rad50-Nbs1 (MRN) complex used to study DNA double-strand break signaling and ATM/ATR activation. It is supplied as a solid and as DMSO solutions, with molecular formula C10H8N2O2S, molecular weight 220.25 g/mol, CAS 299953-00-7, and reported purity ≈99.2%.
  • Inhibits MRN-dependent ATM activation for DNA damage research.
  • Molecular formula C10H8N2O2S; molecular weight 220.25 g/mol.
  • High purity (~99.2%) appropriate for in vitro assays.
  • Available as a 50 MG solid and as 10 mM solutions in DMSO.
  • Useful for biochemical and cell-based studies of DNA repair pathways.
Catalog No. 50-002-26682 Supplier Medchemexpress LLC Supplier No. HY11769350MG
(E/Z)-Mirin is a research-grade small-molecule inhibitor of the Mre11-Rad50-Nbs1 (MRN) complex used to study DNA double-strand break signaling and ATM/ATR activation. It is supplied as a solid and as DMSO solutions, with molecular formula C10H8N2O2S, molecular weight 220.25 g/mol, CAS 299953-00-7, and reported purity ≈99.2%.
  • Inhibits MRN-dependent ATM activation for DNA damage research.
  • Molecular formula C10H8N2O2S; molecular weight 220.25 g/mol.
  • High purity (~99.2%) appropriate for in vitro assays.
  • Available as a 50 MG solid and as 10 mM solutions in DMSO.
  • Useful for biochemical and cell-based studies of DNA repair pathways.