Learn More
Abcam Mirin, MRN-ATM comple x inhibitor, 25MG
Catalog No. 502706826
Encompass_Preferred
MW 220.25 Da, Purity >99%. Mre11-Rad50-Nbs1 (MRN)-ATM complex inhibitor that blocks the 3' to 5' exonuclease activity associated with Mre11. Prevents ATM activation in response to double strand breaks (IC₅₀= 12 μM) and induces G₂ arrest in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6826 Supplier Abcam Supplier No. AB14118225MG
MW 220.25 Da, Purity >99%. Mre11-Rad50-Nbs1 (MRN)-ATM complex inhibitor that blocks the 3' to 5' exonuclease activity associated with Mre11. Prevents ATM activation in response to double strand breaks (IC₅₀= 12 μM) and induces G₂ arrest in vitro.
The product is subject to the following: Abcam Restricted Use Statement