Learn More
Selleck Chemical LLC Miriplatin S5024-25mg
SDP
Supplier:  Selleck Chemical LLC S502425MG
Encompass_Preferred
Miriplatin (SM-11355) is a derivative of cisplatin containing myristates as a carrier ligand It is a novel lipophilic platinum complex developed to treat hepatocellular carcinoma DMSO is not recommended to dissolve platinum-based drugs which can easily lead to drug inactivation
Catalog No. 50-311-0911