Learn More
Medchemexpress LLC Mirodenafil dihydrochloride | 862189-96-6 | MFCD28139019 | 99.9% | 604.59 g/mol | C26H39Cl2N5O5S | 10 MG
SDP
Catalog No. 5000285714
Encompass_Preferred
Mirodenafil dihydrochloride is an orally active, potent, reversible, and selective phosphodiesterase 5 (PDE5) inhibitor with reported glucocorticoid receptor modulatory activity. It has been used in preclinical research to modulate Wnt/β-catenin signaling and is studied for erectile dysfunction, Alzheimer's disease, and systemic sclerosis research applications.
  • Potent and selective PDE5 inhibition for target-specific studies.
  • Reversible inhibitor enabling transient modulation in assays.
  • Modulates glucocorticoid receptor activity relevant to receptor signaling studies.
  • Activates Wnt/β-catenin signaling via Dkk1 downregulation.
  • High purity suitable for biochemical and cellular assays.
  • Available in solid form and as DMSO solution for flexible dosing.
Catalog No. 50-002-85714 Supplier Medchemexpress LLC Supplier No. HY14930A10MG
Mirodenafil dihydrochloride is an orally active, potent, reversible, and selective phosphodiesterase 5 (PDE5) inhibitor with reported glucocorticoid receptor modulatory activity. It has been used in preclinical research to modulate Wnt/β-catenin signaling and is studied for erectile dysfunction, Alzheimer's disease, and systemic sclerosis research applications.
  • Potent and selective PDE5 inhibition for target-specific studies.
  • Reversible inhibitor enabling transient modulation in assays.
  • Modulates glucocorticoid receptor activity relevant to receptor signaling studies.
  • Activates Wnt/β-catenin signaling via Dkk1 downregulation.
  • High purity suitable for biochemical and cellular assays.
  • Available in solid form and as DMSO solution for flexible dosing.