Learn More
Cayman Chemical MIrodenafIlhydrochlorIde 10mg
SDP
Catalog No. 502940117
Encompass_Preferred
A PDE5 inhibitor; increases penile ICP in rat models of STZ-induced diabetes and cavernosal nerve injury at 1 and 10 mg/kg, respectively; decreases bladder wall submucosal fibrosis and degeneration in a rat model of chronic bladder ischemia at 4 mg/kg per day; decreases bladder overactivity in a female rat model of partial bladder outlet obstruction
Catalog No. 50-294-0117 Supplier Cayman Chemical Supplier No. 2904510 MG
A PDE5 inhibitor; increases penile ICP in rat models of STZ-induced diabetes and cavernosal nerve injury at 1 and 10 mg/kg, respectively; decreases bladder wall submucosal fibrosis and degeneration in a rat model of chronic bladder ischemia at 4 mg/kg per day; decreases bladder overactivity in a female rat model of partial bladder outlet obstruction