Learn More
Medchemexpress LLC HY-12650 100mg , Mirogabalin DS5565 CAS:1138245-13-2 Purity:98%
SDP
Catalog No. 501493428
Encompass_Preferred
Medchemexpress, HY-12650 100mg Mirogabalin DS5565 CAS:1138245-13-2 Formula:C12H19NO2 α2δ-1 Calcium Channel Purity:98% Mirogabalin (DS-5565) is a novel, preferentially selective α2δ-1 ligand characterized by high potency and selectivity to the α2δ-1 subunit of voltage-sensitive calcium channel complexes in the CNS. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3428 Supplier Medchemexpress LLC Supplier No. HY12650100MG
Medchemexpress, HY-12650 100mg Mirogabalin DS5565 CAS:1138245-13-2 Formula:C12H19NO2 α2δ-1 Calcium Channel Purity:98% Mirogabalin (DS-5565) is a novel, preferentially selective α2δ-1 ligand characterized by high potency and selectivity to the α2δ-1 subunit of voltage-sensitive calcium channel complexes in the CNS. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.