Learn More
Medchemexpress LLC Mirogabalin besylate | 1138245-21-2 | 99.6% | 367.46 g·mol⁻¹ | C18H25NO5S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY108006100MG
Encompass_Preferred
Mirogabalin besylate is a research-grade small molecule ligand for the α2δ subunit of voltage-gated calcium channels, provided as a solid and in solution formats for in vitro and preclinical studies. It is supplied with supporting documents (COA, SDS) and is intended for laboratory research use.
  • Selective α2δ subunit ligand for calcium channels.
  • High reported purity suitable for biochemical assays.
  • White to off-white solid, supplied in multiple package sizes.
  • Available as 10 mM solution in DMSO for ready-to-use assays.
  • Certificate of analysis and safety data sheet available.
  • Store sealed at 4°C; in solvent store at -80°C or -20°C as specified.
Catalog No. 50-002-42253