Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mirogabalin besylate | 1138245-21-2 | MFCD31631215 | 99.6% | 367.46 | C18H25NO5S | 10 MG
SDP

Catalog No. 5000241944
Encompass_Preferred

Mirogabalin besylate is the besylate salt of a selective, orally available ligand for the α2δ subunit of voltage-gated calcium channels, provided for research use with manufacturer-specified purity, solubility, and storage recommendations.

  • Selective α2δ ligand with low-nanomolar binding affinity to human α2δ-1 and α2δ-2.
  • High purity: 99.57% (manufacturer stated).
  • White to off-white solid suitable for analytical and preparative workflows.
  • Available as small research packs and as DMSO solutions ready for use.
  • Soluble in DMSO at 100 mg/mL; may require sonication and use of fresh DMSO.
  • Storage: solid at 4°C sealed, solutions at -20°C to -80°C per stability guidance.
  • For research use only; not for human or veterinary use and subject to territorial restrictions.

Catalog No. 50-002-41944 Supplier Medchemexpress LLC Supplier No. HY10800610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mirogabalin besylate is the besylate salt of a selective, orally available ligand for the α2δ subunit of voltage-gated calcium channels, provided for research use with manufacturer-specified purity, solubility, and storage recommendations.

  • Selective α2δ ligand with low-nanomolar binding affinity to human α2δ-1 and α2δ-2.
  • High purity: 99.57% (manufacturer stated).
  • White to off-white solid suitable for analytical and preparative workflows.
  • Available as small research packs and as DMSO solutions ready for use.
  • Soluble in DMSO at 100 mg/mL; may require sonication and use of fresh DMSO.
  • Storage: solid at 4°C sealed, solutions at -20°C to -80°C per stability guidance.
  • For research use only; not for human or veterinary use and subject to territorial restrictions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.