Learn More
Medchemexpress LLC Mirogabalin besylate | 1138245-21-2 | MFCD31631215 | 99.6% | 367.46 | C18H25NO5S | 50 MG
SDP
Catalog No. 5000242234
Encompass_Preferred
Mirogabalin besylate is a research-grade small molecule that acts as a selective, orally available ligand for the α2δ subunit of voltage-gated calcium channels. It is supplied as a high-purity solid with manufacturer documentation supporting identity, purity, and safe handling for laboratory research.
  • High purity (99.6%) suitable for research applications.
  • Molecular formula C18H25NO5S; molecular weight 367.46.
  • Solid powder formulation supplied in a 50 MG unit.
  • Store solid sealed at 4°C, away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-42234 Supplier Medchemexpress LLC Supplier No. HY10800650MG
Mirogabalin besylate is a research-grade small molecule that acts as a selective, orally available ligand for the α2δ subunit of voltage-gated calcium channels. It is supplied as a high-purity solid with manufacturer documentation supporting identity, purity, and safe handling for laboratory research.
  • High purity (99.6%) suitable for research applications.
  • Molecular formula C18H25NO5S; molecular weight 367.46.
  • Solid powder formulation supplied in a 50 MG unit.
  • Store solid sealed at 4°C, away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.