Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mirogabalin besylate | 1138245-21-2 | MFCD31631215 | 99.6% | 367.46 g·mol⁻¹ | C18H25NO5S | 1 ML
SDP

Catalog No. 5000241791
Encompass_Preferred

Mirogabalin besylate is the besylate salt of mirogabalin, a selective ligand for the α2δ subunit of voltage-gated calcium channels used in preclinical research. Supplied as a ready-to-use solution in DMSO, it is intended for in vitro and related laboratory applications. Refer to the safety data sheet for handling and storage instructions.

  • 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.6%) as supplied.
  • Molecular weight 367.46 g·mol⁻¹.
  • Molecular formula C18H25NO5S.
  • Recommended storage: in solvent -80°C (up to 6 months) or -20°C (up to 1 month); solid 4°C (sealed).
  • Intended for research use only.

Catalog No. 50-002-41791 Supplier Medchemexpress LLC Supplier No. HY10800610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mirogabalin besylate is the besylate salt of mirogabalin, a selective ligand for the α2δ subunit of voltage-gated calcium channels used in preclinical research. Supplied as a ready-to-use solution in DMSO, it is intended for in vitro and related laboratory applications. Refer to the safety data sheet for handling and storage instructions.

  • 10 mM solution in DMSO, 1 mL vial.
  • High purity (99.6%) as supplied.
  • Molecular weight 367.46 g·mol⁻¹.
  • Molecular formula C18H25NO5S.
  • Recommended storage: in solvent -80°C (up to 6 months) or -20°C (up to 1 month); solid 4°C (sealed).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.