Learn More
World Precision Instrument Mirror Microinjection Base with 76 mm Focus Mount
SDP
Catalog No. 502539226
Encompass
PZMIII-MI Microscope with Illuminated Base and Articulating Mirror is perfect for microinjection and transfection. It includes our standard stereo microscope head mounted on a research grade Brightfield/Darkfield pole type stand. It has a large stable work surface and a rotatable lens/mirror system which provides transmitted LED intensity illumination.
The sliding mirror is gimbaled, allowing for a full range of movement front to back, as well as rotation. The mirror rotates 360º on one axis and can slide for further lighting effect directionally, front to back. A knob on the right of the base adjusts the mirror and a locking ring holds the desired mirror position.
The articulating mirror lets you vary the microscope illumination from Brightfield LED to Darkfield LED at an appropriate angle. It is an effective tool for viewing live bacteria.
Catalog No. 50-253-9226 Supplier World Precision Instrument Supplier No. 505145
PZMIII-MI Microscope with Illuminated Base and Articulating Mirror is perfect for microinjection and transfection. It includes our standard stereo microscope head mounted on a research grade Brightfield/Darkfield pole type stand. It has a large stable work surface and a rotatable lens/mirror system which provides transmitted LED intensity illumination.
The sliding mirror is gimbaled, allowing for a full range of movement front to back, as well as rotation. The mirror rotates 360º on one axis and can slide for further lighting effect directionally, front to back. A knob on the right of the base adjusts the mirror and a locking ring holds the desired mirror position.
The articulating mirror lets you vary the microscope illumination from Brightfield LED to Darkfield LED at an appropriate angle. It is an effective tool for viewing live bacteria.