Learn More
Vernier™ Mirror Set for Optics Expansion Kit
SDPSCI_ED
Catalog No. S16209ND Shop All Vernier Software Products
Change view
Click to view available options
Includes: Fixed Convex Mirror, Half Screen, Adjustable Concave Mirror
Catalog No.
S16209ND Includes: Fixed Convex Mirror, Half Screen, Adjustable Concave Mirror
1 options
Catalog No. S16209ND Supplier Vernier™ Supplier No. MOEK
Add to Cart
Add to Cart
Includes
Fixed Convex Mirror (-200 mm focal length), Half Screen, Adjustable Concave Mirror (200 mm focal length)
The Mirror Set for the Optics Expansion Kit extends the kit to allow students to easily study image formation by concave and convex mirrors.
  • Includes a Half Screen, an adjustable Concave Mirror, and a fixed Convex Mirror.
  • Use these parts with the Light Source and Screen on the Optics Expansion Kit and the track from the Dynamics System to observe and measure real and virtual images cast by mirrors.
  • With mirrors solidly mounted on a track acting as an optics bench, your students can quickly understand how images are formed and located.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Educational use only
Includes Fixed Convex Mirror, Half Screen, Adjustable Concave Mirror
Length (Metric) 300 mm focal length (adjustable concave mirror), -200 mm focal length (fixed convex mirror)