Learn More
Electron Microscopy Sciences Mirror Stereo Viewer
SDP
Catalog No. 501899207
Encompass_Preferred
For viewing 4"x5" stereo pairs. Magnifier can be replaced. Base line spacing: 260mm, magnification: 1.8X, field of view without magnifier: 260x200mm, field of view with magnifier: 210x150mm, maximum size set up: 713x2.
Catalog No. 50-189-9207 Supplier Electron Microscopy Sciences Supplier No. 68105
For viewing 4"x5" stereo pairs. Magnifier can be replaced. Base line spacing: 260mm, magnification: 1.8X, field of view without magnifier: 260x200mm, field of view with magnifier: 210x150mm, maximum size set up: 713x2.