Learn More
Electron Microscopy Sciences Mirsky's Fixative Ready-to-Use 2 LT
SDP
Supplier:  Electron Microscopy Sciences 1556120
Encompass_Preferred
Saponin/Glyoxylate fixative is for use in immunohistological and immunological protocols. It contains no formaldehyde or glutaraldehyde giving it reduced toxicity and virtually no odor. Neutral, buffered, and isotonic (308 mOsmol). Additionally, Mirsky's Fixative does not contain toxic or hazardous buffers such as cacodylate or barbital.
Double reactive sites afford excellent cross-linking properties while maintaining sample enzyme activity. Samples processed in Mirsky's Fixative for light microscopy can subsequently be used in electron microscopy procedures.
Catalog No. 50-190-1208