Learn More
Cayman Chemical Mirtazapine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1216678-68-0, 1 mg
SDP
Catalog No. 502978399
Encompass_Preferred
An internal standard for the quantification of mirtazapine by GC- or LC-MS
Catalog No. 50-297-8399 Supplier Cayman Chemical Supplier No. 401211 MG
An internal standard for the quantification of mirtazapine by GC- or LC-MS