Learn More
Medchemexpress LLC Mirtazapine | 85650-52-8 | 99.9% | 50 MG
SDP
Catalog No. 5000406610
Encompass_Preferred
Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It acts as a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist.
  • Potent and orally active noradrenergic and specific serotonergic antidepressant.
  • Antagonizes adrenergic α2-autoreceptors and α2-heteroreceptors, and blocks 5-HT2 and 5-HT3 receptors.
  • Enhances the release of norepinephrine and 5-HT1A-mediated serotonergic transmission.
  • Significantly reduces activation-induced release of cytokine/chemokine mediators from human CD14+ monocytes in vitro.
  • Inhibits hepatic innate immune networks to attenuate immune-mediated liver injury in mice in vivo.
  • Suppresses Con A-induced increases in hepatic ICAM-1 expression, reducing neutrophil recruitment into the liver.
Catalog No. 50-004-06610 Supplier Medchemexpress LLC Supplier No. HYB035250MG
Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It acts as a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist.
  • Potent and orally active noradrenergic and specific serotonergic antidepressant.
  • Antagonizes adrenergic α2-autoreceptors and α2-heteroreceptors, and blocks 5-HT2 and 5-HT3 receptors.
  • Enhances the release of norepinephrine and 5-HT1A-mediated serotonergic transmission.
  • Significantly reduces activation-induced release of cytokine/chemokine mediators from human CD14+ monocytes in vitro.
  • Inhibits hepatic innate immune networks to attenuate immune-mediated liver injury in mice in vivo.
  • Suppresses Con A-induced increases in hepatic ICAM-1 expression, reducing neutrophil recruitment into the liver.