Learn More
Medchemexpress LLC Mirtazapine | 85650-52-8 | 99.9% | 500 MG
SDP
Catalog No. 5000405315
Encompass_Preferred
Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It functions as a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist, influencing norepinephrine and 5-HT1A-mediated serotonergic transmission. It is primarily metabolized by CYP1A2, CYP2D6, and CYP3A4.
  • Potent and orally active antidepressant
  • Acts as a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist
  • Enhances release of norepinephrine
  • Enhances 5-HT1A-mediated serotonergic transmission
  • Reduces activation-induced release of cytokine/chemo kine mediators from human CD14+ monocytes in vitro
  • Inhibits Con A-induced liver injury in vivo by suppressing hepatic macrophage/monocyte activation and pro-inflammatory cytokine production
Catalog No. 50-004-05315 Supplier Medchemexpress LLC Supplier No. HYB0352500MG
Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It functions as a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist, influencing norepinephrine and 5-HT1A-mediated serotonergic transmission. It is primarily metabolized by CYP1A2, CYP2D6, and CYP3A4.
  • Potent and orally active antidepressant
  • Acts as a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist
  • Enhances release of norepinephrine
  • Enhances 5-HT1A-mediated serotonergic transmission
  • Reduces activation-induced release of cytokine/chemo kine mediators from human CD14+ monocytes in vitro
  • Inhibits Con A-induced liver injury in vivo by suppressing hepatic macrophage/monocyte activation and pro-inflammatory cytokine production