Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mirtazapine | 85650-52-8 | MFCD00865427 | 99.9% | 25 MG
SDP

Catalog No. 5000450632
Encompass_Preferred

Mirtazapine is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It acts as an antagonist for 5-HT2, 5-HT3, histamine H1 receptors, and α2-adrenoceptors. This compound is intended for research use only and is not sold to patients.

  • Potent and orally active NaSSA agent
  • Antagonist of multiple receptors: 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor
  • Enhances norepinephrine and 5-HT1A-mediated serotonergic transmission
  • Inhibits hepatic macrophage/monocyte activation, decreases pro-inflammatory cytokine and chemokine production, and suppresses ICAM-1 expression, leading to reduced neutrophil recruitment into the liver (observed in mice)
  • Metabolized by CYP1A2, CYP2D6, and CYP3A4 isoenzymes
  • Significantly reduces activation-induced release of cytokine/chemokine mediators from human CD14+ monocytes in vitro

Catalog No. 50-004-50632 Supplier Medchemexpress LLC Supplier No. 25MGHYB0352R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mirtazapine is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It acts as an antagonist for 5-HT2, 5-HT3, histamine H1 receptors, and α2-adrenoceptors. This compound is intended for research use only and is not sold to patients.

  • Potent and orally active NaSSA agent
  • Antagonist of multiple receptors: 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor
  • Enhances norepinephrine and 5-HT1A-mediated serotonergic transmission
  • Inhibits hepatic macrophage/monocyte activation, decreases pro-inflammatory cytokine and chemokine production, and suppresses ICAM-1 expression, leading to reduced neutrophil recruitment into the liver (observed in mice)
  • Metabolized by CYP1A2, CYP2D6, and CYP3A4 isoenzymes
  • Significantly reduces activation-induced release of cytokine/chemokine mediators from human CD14+ monocytes in vitro

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.