Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mirtazapine | 85650-52-8 | MFCD00865427 | 99.9% | 265.36 | C17H19N3 | 10 MG
SDP

Catalog No. 5000298967
Encompass_Preferred

Mirtazapine is a tetracyclic antidepressant used in research to probe noradrenergic and specific serotonergic mechanisms. This research-grade 10 mg sample is supplied as a solid for laboratory use and is characterized by its reported high purity and defined molecular properties.

  • Potent NaSSA agent for studies of noradrenergic and serotonergic pathways.
  • Antagonist of 5-HT2, 5-HT3, H1, and α2 receptors.
  • High reported purity (99.9%) suitable for analytical and in vitro assays.
  • Molecular formula C17H19N3 and molecular weight 265.36 aid compound identification.
  • Primarily metabolized by CYP1A2, CYP2D6, and CYP3A4, relevant for metabolism studies.

Catalog No. 50-002-98967 Supplier Medchemexpress LLC Supplier No. HYB035210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mirtazapine is a tetracyclic antidepressant used in research to probe noradrenergic and specific serotonergic mechanisms. This research-grade 10 mg sample is supplied as a solid for laboratory use and is characterized by its reported high purity and defined molecular properties.

  • Potent NaSSA agent for studies of noradrenergic and serotonergic pathways.
  • Antagonist of 5-HT2, 5-HT3, H1, and α2 receptors.
  • High reported purity (99.9%) suitable for analytical and in vitro assays.
  • Molecular formula C17H19N3 and molecular weight 265.36 aid compound identification.
  • Primarily metabolized by CYP1A2, CYP2D6, and CYP3A4, relevant for metabolism studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.