Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mirtazapine | 85650-52-8 | MFCD00865427 | 99.95% | 100 MG
SDP

Catalog No. 5000407309
Encompass_Preferred

Mirtazapine (Standard) is the analytical standard of Mirtazapine, intended for research and analytical applications. It is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. Mirtazapine is also a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist with pKi values of 8.05, 8.1, 9.3, and 6.95, respectively.

  • Analytical standard for research and analytical applications
  • Potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent
  • 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS

Catalog No. 50-004-07309 Supplier Medchemexpress LLC Supplier No. HYB0352R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mirtazapine (Standard) is the analytical standard of Mirtazapine, intended for research and analytical applications. It is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. Mirtazapine is also a 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist with pKi values of 8.05, 8.1, 9.3, and 6.95, respectively.

  • Analytical standard for research and analytical applications
  • Potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent
  • 5-HT2, 5-HT3, histamine H1 receptor, and α2-adrenoceptor antagonist
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.