Learn More
Medchemexpress LLC Mirtazapine-d3 | 1216678-68-0 | 99.7% | 1 ML
SDP
Catalog No. 5000406283
Encompass_Preferred
Mirtazapine-d3 is a deuterium labeled Mirtazapine, which is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It functions by blocking 5-HT2 and 5-HT3 receptors, and is intended for research use only.
  • Deuterium labeled Mirtazapine
  • 5-HT receptor inhibitor
  • Potent noradrenergic and specific serotonergic antidepressant (NaSSA) agent
  • Blocks 5-HT2 and 5-HT3 receptors
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-004-06283 Supplier Medchemexpress LLC Supplier No. HYB0352S10MM/1ML
Mirtazapine-d3 is a deuterium labeled Mirtazapine, which is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. It functions by blocking 5-HT2 and 5-HT3 receptors, and is intended for research use only.
  • Deuterium labeled Mirtazapine
  • 5-HT receptor inhibitor
  • Potent noradrenergic and specific serotonergic antidepressant (NaSSA) agent
  • Blocks 5-HT2 and 5-HT3 receptors
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)