Learn More
Medchemexpress LLC Mirtazapine-d3 | 1216678-68-0 | M002686 | 99.68% | 5 MG
SDP
Catalog No. 5000406045
Encompass_Preferred
Mirtazapine-d3 is a deuterium-labeled Mirtazapine. Mirtazapine is a 5-HT receptor inhibitor, and a potent, orally active noradrenergic and specific serotonergic antidepressant (NaSSA) that works by blocking 5-HT2 and 5-HT3 receptors.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent, orally active noradrenergic and specific serotonergic antidepressant (NaSSA)
  • Blocks 5-HT2 and 5-HT3 receptors
  • Deuterium-labeled compound
Catalog No. 50-004-06045 Supplier Medchemexpress LLC Supplier No. HYB0352S5MG
Mirtazapine-d3 is a deuterium-labeled Mirtazapine. Mirtazapine is a 5-HT receptor inhibitor, and a potent, orally active noradrenergic and specific serotonergic antidepressant (NaSSA) that works by blocking 5-HT2 and 5-HT3 receptors.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent, orally active noradrenergic and specific serotonergic antidepressant (NaSSA)
  • Blocks 5-HT2 and 5-HT3 receptors
  • Deuterium-labeled compound