Learn More
Selleck Chemical LLC Mirvetuximab-MMAE-D4060-1MG
SDP
Catalog No. 503134763
Encompass_Preferred
Mirvetuximab-MMAE is an ADC targeting FOLR1 (folate receptor 1) It can be used in ovarian and other FR -positive cancer research MW 145 96 KD
Catalog No. 50-313-4763 Supplier Selleck Chemical LLC Supplier No. D40601MG
Mirvetuximab-MMAE is an ADC targeting FOLR1 (folate receptor 1) It can be used in ovarian and other FR -positive cancer research MW 145 96 KD