Learn More
Cell Signaling Technology Mismatch DNA Repair Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502730276
Encompass
The Mismatch DNA Repair (MMR) Antibody Sampler Kit provides an economical means of detecting proteins involved in DNA mismatch repair, a critical process for maintaining genomic integrity. The kit contains a selection of antibodies against key MMR proteins to facilitate comprehensive analysis via western blot, immunoprecipitation, and immunohistochemistry applications.
  • Includes antibodies targeting MLH1, PMS2, MSH2, MSH6, and EXO1
  • Supplied with HRP-linked secondary antibody for western blotting
  • Validated for multiple applications including western blotting, immunoprecipitation, and immunohistochemistry
  • Provided in a stable storage buffer containing glycerol for convenient use at -20°C
Catalog No. 50-273-0276 Supplier Cell Signaling Technology Supplier No. 89884T
The Mismatch DNA Repair (MMR) Antibody Sampler Kit provides an economical means of detecting proteins involved in DNA mismatch repair, a critical process for maintaining genomic integrity. The kit contains a selection of antibodies against key MMR proteins to facilitate comprehensive analysis via western blot, immunoprecipitation, and immunohistochemistry applications.
  • Includes antibodies targeting MLH1, PMS2, MSH2, MSH6, and EXO1
  • Supplied with HRP-linked secondary antibody for western blotting
  • Validated for multiple applications including western blotting, immunoprecipitation, and immunohistochemistry
  • Provided in a stable storage buffer containing glycerol for convenient use at -20°C