Learn More
Cayman Chemical Misonidazole, Purity: ≥95%, CAS: 13551-87-6, 10 mg
SDP
Catalog No. 502973268
Encompass_Preferred
A nitroimidazole with radiosensitizing and antineoplastic properties; metabolites accumulate in hypoxic cells and can themselves be cytotoxic or they can increase the chemosensitivity and radiosensitivity of the target cells; used in the imaging of hypoxic regions in tumors and in the cardiovascular system
Catalog No. 50-297-3268 Supplier Cayman Chemical Supplier No. 1560610 MG
A nitroimidazole with radiosensitizing and antineoplastic properties; metabolites accumulate in hypoxic cells and can themselves be cytotoxic or they can increase the chemosensitivity and radiosensitivity of the target cells; used in the imaging of hypoxic regions in tumors and in the cardiovascular system