Learn More
Cayman Chemical Misoprostold5 GC MS Standards
SDP
Supplier:  Cayman Chemical 10216100 UG
Encompass_Preferred
An internal standard for the quantification of misoprostol by GC- or LC-MS
Catalog No. 50-288-8563