Learn More
Cayman Chemical Misoprostol-d5, Purity: ≥99% deuterated forms (d1-d5), 1 mg
SDP
Catalog No. 502889115
Encompass_Preferred
An internal standard for the quantification of misoprostol by GC- or LC-MS
Catalog No. 50-288-9115 Supplier Cayman Chemical Supplier No. 102161 MG
An internal standard for the quantification of misoprostol by GC- or LC-MS