Learn More
Cayman Chemical Misoprostold5 GC MS Standards
SDP
Catalog No. 502888343
Encompass_Preferred
An internal standard for the quantification of misoprostol by GC- or LC-MS
Catalog No. 50-288-8343 Supplier Cayman Chemical Supplier No. 1021650 UG
An internal standard for the quantification of misoprostol by GC- or LC-MS