Learn More
Cayman Chemical Misoprostol (free acid), Purity: ≥97%, CAS: 112137-89-0, 50 mg
SDP
Catalog No. 502955278
Encompass_Preferred
Misoprostol is a prostaglandin E1 (Item No. 13010) analog with agonist activity mediated by EP2, EP3, and EP4 receptors. It has been shown to inhibit the formation of gastric lesions in rats (ED50 = 0.31 µg/kg),{1754} inhibit superoxide generation in human neutrophils (EC50 = 0.35 µM), and relax fetal rabbit ductus arteriosus (EC50 = 0.36 nM) in a concentration dependent manner. Misoprostol is commonly used in clinical medicine for the prevention of peptic ulcer disease. It has also been used in conjunction with mifepristone (RU-486; Item No. 10006317) for the oral induction of first trimester abortion. Misoprostol contains a C-1 methyl ester and is readily absorbed and rapidly hydrolyzed in humans to the active free acid.{2313}
Catalog No. 50-295-5278 Supplier Cayman Chemical Supplier No. 1382150 MG
Misoprostol is a prostaglandin E1 (Item No. 13010) analog with agonist activity mediated by EP2, EP3, and EP4 receptors. It has been shown to inhibit the formation of gastric lesions in rats (ED50 = 0.31 µg/kg),{1754} inhibit superoxide generation in human neutrophils (EC50 = 0.35 µM), and relax fetal rabbit ductus arteriosus (EC50 = 0.36 nM) in a concentration dependent manner. Misoprostol is commonly used in clinical medicine for the prevention of peptic ulcer disease. It has also been used in conjunction with mifepristone (RU-486; Item No. 10006317) for the oral induction of first trimester abortion. Misoprostol contains a C-1 methyl ester and is readily absorbed and rapidly hydrolyzed in humans to the active free acid.{2313}