Learn More
ACROBiosystems ACROBiosystems Human MIS RII Protein, Fc Tag
Supplier:  ACROBiosystems MIIH52581MG
Encompass_Preferred
Recombinant Protein;1MG;Human MIS RII, Fc Tag (MII-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - Ser 144 (Accession # Q16671-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.0 kDa.
Catalog No. 50-195-6699