Learn More
Diamond Diagnostics Mission Complete Linearity Controls, 4 vials of each of 5 levels at 1.8mL.
SDP
Catalog No. NC0965615
Encompass
Mission Complete Linearity Control contains 4 vials of each of 5 levels at 1.8mL. For use on all Blood Gas and Electrolyte Systems, assayed for pH, pCO2, pO2, Na+, K+, Ca++, Cl-, Li+ Glucose and Lactate. Assayed for Abbott, Diamond, Idexx, Instrumentation Laboratory, Medica, Nova Biomedical, Optimedical, Radiometer, Roche Diangostics and Siemens. Compatible with: Medica Easylyte series , Diamond ProLyte , IL ILyte and Menarini Spotlyte , Alfa Wassermann Starlyte III, Starlyte V , Diamond SmartLyte , Roche 9110, 9120, 9130, 9140, 9180, 9181 , Radiometer ABL 5, 50, 500, 510, 520, 555, EML 100, 105 , Siemens Rapidlab 840, 845, 850, 855, 860, 865 , Diamond CareLyte , MISSION CONTROL Blood Gas, Electrolyte, Metabolite & Cooximetry Controls , Roche AVL Compact 1-2-3 , Siemens Rapidlab 248, 348
Catalog No. NC0965615 Supplier Diamond Diagnostics Supplier No. DD92900
Mission Complete Linearity Control contains 4 vials of each of 5 levels at 1.8mL. For use on all Blood Gas and Electrolyte Systems, assayed for pH, pCO2, pO2, Na+, K+, Ca++, Cl-, Li+ Glucose and Lactate. Assayed for Abbott, Diamond, Idexx, Instrumentation Laboratory, Medica, Nova Biomedical, Optimedical, Radiometer, Roche Diangostics and Siemens. Compatible with: Medica Easylyte series , Diamond ProLyte , IL ILyte and Menarini Spotlyte , Alfa Wassermann Starlyte III, Starlyte V , Diamond SmartLyte , Roche 9110, 9120, 9130, 9140, 9180, 9181 , Radiometer ABL 5, 50, 500, 510, 520, 555, EML 100, 105 , Siemens Rapidlab 840, 845, 850, 855, 860, 865 , Diamond CareLyte , MISSION CONTROL Blood Gas, Electrolyte, Metabolite & Cooximetry Controls , Roche AVL Compact 1-2-3 , Siemens Rapidlab 248, 348