Learn More
Chemglass Life Sciences Mistral Sample Concentrator, Without Needle Head, Porvair, 110V
SDP
Catalog No. 501941139
Encompass
Mistral Sample Concentrator, Without Needle Head, Porvair, 110V. The Ultravap Mistral from Porvair Sciences is designed to remove the traditional laboratory ‘bottleneck’ of solvent evaporation from microplates. Fully automating the dry-down step has always been impossible because it is difficult to interface liquid handling robots with traditional centrifugal-type evaporators. The Ultravap Mistral design, by dispensing with the rotating arm of the centrifuge, overcomes this problem and is much better positioned to link with your robot. With more than 20 years’ experience in producing deep well microplates, Porvair Sciences has thoroughly researched the problems of drying down organic solvents in plates. This has led to the ultimate microplate blow down evaporator – the Ultravap Mistral.
Catalog No. 50-194-1139 Supplier Chemglass Life Sciences Supplier No. CLS3599C100
Add to Cart
Add to Cart
Request a Quote
Mistral Sample Concentrator, Without Needle Head, Porvair, 110V. The Ultravap Mistral from Porvair Sciences is designed to remove the traditional laboratory ‘bottleneck’ of solvent evaporation from microplates. Fully automating the dry-down step has always been impossible because it is difficult to interface liquid handling robots with traditional centrifugal-type evaporators. The Ultravap Mistral design, by dispensing with the rotating arm of the centrifuge, overcomes this problem and is much better positioned to link with your robot. With more than 20 years’ experience in producing deep well microplates, Porvair Sciences has thoroughly researched the problems of drying down organic solvents in plates. This has led to the ultimate microplate blow down evaporator – the Ultravap Mistral.