Learn More
MSC Delfin Industrial Vacuum Cleaner, MTL 202 DS, Single-Phase, 1 ea
Catalog No. 502816129
Encompass
The MTL 202 DS industrial vacuum cleaner features two single-phase motors and a large surface area class M filter, offering extreme maneuverability. It is an ideal suction solution for laboratories or production plants, ensuring maximum performance with its DUSTOP system for filter cleaning by differential pressure.
  • Two single-phase motors
  • Class M PTFE primary filter with Dustop cleaning system
  • Integrated cyclone
  • Compact and mobile footprint
  • Painted steel construction
  • Bin capacity of 20 L
  • Maximum vacuum rate of 2,500
  • Maximum airflow of 360
  • Voltage of 230 V, 50 Hz
Catalog No. 50-281-6129 Supplier MSC Supplier No. 13677448
Add to Cart
Add to Cart
Request a Quote
The MTL 202 DS industrial vacuum cleaner features two single-phase motors and a large surface area class M filter, offering extreme maneuverability. It is an ideal suction solution for laboratories or production plants, ensuring maximum performance with its DUSTOP system for filter cleaning by differential pressure.
  • Two single-phase motors
  • Class M PTFE primary filter with Dustop cleaning system
  • Integrated cyclone
  • Compact and mobile footprint
  • Painted steel construction
  • Bin capacity of 20 L
  • Maximum vacuum rate of 2,500
  • Maximum airflow of 360
  • Voltage of 230 V, 50 Hz