Learn More
MSC Delfin Industrial Mistral 202 DS Vacuum, 5 gal, 1 ea
Catalog No. 502817166
Encompass
The Mistral 202 DS is a versatile industrial vacuum system designed for efficient dust removal in various laboratory and production plant applications. This compact and portable machine features two single-phase motors and a Class M Polyester PTFE filter, offering efficient cleaning and dust extraction.
  • Suitable for dry and solids applications
  • Compact and portable design for easy mobility
  • Constructed with powder-coated steel
  • Equipped with two powerful bypass motors with independent cooling
  • Features a roll-out collection container and optional disposable bags
  • Includes a Class M Polyester PTFE filter with indicator light
  • Tangential inlet with internal cyclone design
  • Utilizes a DUSTOP cleaning system with pressure differential
Catalog No. 50-281-7166 Supplier MSC Supplier No. 28835742
Add to Cart
Add to Cart
Request a Quote
The Mistral 202 DS is a versatile industrial vacuum system designed for efficient dust removal in various laboratory and production plant applications. This compact and portable machine features two single-phase motors and a Class M Polyester PTFE filter, offering efficient cleaning and dust extraction.
  • Suitable for dry and solids applications
  • Compact and portable design for easy mobility
  • Constructed with powder-coated steel
  • Equipped with two powerful bypass motors with independent cooling
  • Features a roll-out collection container and optional disposable bags
  • Includes a Class M Polyester PTFE filter with indicator light
  • Tangential inlet with internal cyclone design
  • Utilizes a DUSTOP cleaning system with pressure differential