Learn More
MSC Delfin Industrial Vacuum, TECNOIL TC 100 MPI, 1 ea
Catalog No. 502819675
Encompass
The TECNOIL TC 100 MPI is a powerful single-phase industrial vacuum cleaner engineered for the mechanical industry and CNC machine tool maintenance. It collects coolant liquids and emulsions, simultaneously separating metal chips to enable liquid reuse, resulting in significant savings in cost and servicing time.
  • Collects coolant liquids and emulsions, separating metal chips for reuse
  • Provides 26 gal (100 L) liquid capacity and 10.6 gal (40 L) solids capacity
  • Simultaneous collection and rapid discharge via independent submerged pump
  • Compact, portable design constructed with powder-coated steel
  • Two powerful bypass motors with independent cooling
  • Camlock connection on hose for improved liquid discharge sealing
  • Includes oil-proof wheels, gaskets, and a level detector
  • Reduces downtime, disposal, and maintenance costs
Catalog No. 50-281-9675 Supplier MSC Supplier No. 13677018
The TECNOIL TC 100 MPI is a powerful single-phase industrial vacuum cleaner engineered for the mechanical industry and CNC machine tool maintenance. It collects coolant liquids and emulsions, simultaneously separating metal chips to enable liquid reuse, resulting in significant savings in cost and servicing time.
  • Collects coolant liquids and emulsions, separating metal chips for reuse
  • Provides 26 gal (100 L) liquid capacity and 10.6 gal (40 L) solids capacity
  • Simultaneous collection and rapid discharge via independent submerged pump
  • Compact, portable design constructed with powder-coated steel
  • Two powerful bypass motors with independent cooling
  • Camlock connection on hose for improved liquid discharge sealing
  • Includes oil-proof wheels, gaskets, and a level detector
  • Reduces downtime, disposal, and maintenance costs