Learn More
MSC Misty Carpet Cleaner Defoamer, Water-based Silicone, Unscented, 1 gal Bottle
Catalog No. 502836603
Encompass
This carpet cleaner defoamer is a liquid defoamer designed for carpet and upholstery cleaning systems. It is effective for facilities and cleaning teams that utilize steam cleaners, commercial extractors, automatic scrubbers, or wet pick-up vacuums to suppress foam during cleaning. This product features an unscented, water-based silicone formula and is packaged in 1-gallon bottles, with 4 bottles per carton.
  • Water-based silicone formula helps suppress foam.
  • Eliminates potential for sticky residues.
  • Suitable for use in all types of aqueous systems, including steam cleaners, commercial extractors, automatic scrubbers, and wet pick-up vacuums.
  • Unscented.
  • Works in hot or cold aqueous systems.
  • Concentrated liquid formula with variable dilution.
Catalog No. 50-283-6603 Supplier MSC Supplier No. 50539246
This carpet cleaner defoamer is a liquid defoamer designed for carpet and upholstery cleaning systems. It is effective for facilities and cleaning teams that utilize steam cleaners, commercial extractors, automatic scrubbers, or wet pick-up vacuums to suppress foam during cleaning. This product features an unscented, water-based silicone formula and is packaged in 1-gallon bottles, with 4 bottles per carton.
  • Water-based silicone formula helps suppress foam.
  • Eliminates potential for sticky residues.
  • Suitable for use in all types of aqueous systems, including steam cleaners, commercial extractors, automatic scrubbers, and wet pick-up vacuums.
  • Unscented.
  • Works in hot or cold aqueous systems.
  • Concentrated liquid formula with variable dilution.